Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Otoa
Entrez Gene ID
246190
Gene Name
otoancorin
Gene Aliases
-
Chromosome Location
Chr.7: 121083438 - 121163470 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139310.1 NP_647471.1 18 - 19 1962 95
GenBank mRNA AK157904.1 - 18 - 19 1994 95
AK158280.1 - 8 - 9 935 95
AK158326.1 - 8 - 9 931 95
AY055122.1 - 18 - 19 1962 95

View Full Product Details