The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144828.1 | NP_659077.1 | 5 - 6 | 779 | 57 |
GenBank mRNA | AB221544.1 | - | 5 - 6 | 428 | 57 |
AF281662.1 | - | 1 - 2 | 186 | 57 | |
AK141563.1 | - | 3 - 4 | 304 | 57 | |
AY640620.1 | - | 5 - 6 | 457 | 57 | |
AY640621.1 | - | 4 - 5 | 915 | 57 | |
BC011122.1 | - | 5 - 6 | 689 | 57 | |
BC026568.1 | - | 5 - 6 | 779 | 57 | |
BC031129.1 | - | 5 - 6 | 527 | 57 |