Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nek9
Entrez Gene ID
217718
Gene Name
NIMA (never in mitosis gene a)-related expressed kinase 9
Gene Aliases
C130021H08Rik
Chromosome Location
Chr.12: 85299514 - 85339362 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145138.2 NP_660120.2 4 - 5 661 138
GenBank mRNA AJ489828.1 - 4 - 5 636 138
AK173323.1 - 4 - 5 645 138
BC117971.2 - 4 - 5 602 138
BC117972.2 - 4 - 5 601 138

View Full Product Details