The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145853.2 | NP_665852.1 | 27 - 28 | 2576 | 60 |
GenBank mRNA | AF217002.1 | - | 27 - 28 | 2612 | 60 |
AK129303.1 | - | 22 - 23 | 1685 | 60 | |
AK137626.1 | - | 25 - 26 | 2274 | 60 | |
AK148129.1 | - | 27 - 28 | 2637 | 60 | |
BC058951.1 | - | 27 - 28 | 2576 | 60 |