Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspear
Entrez Gene ID
252974
Gene Name
thrombospondin type laminin G domain and EAR repeats
Gene Aliases
C330046G03Rik, ORF65, TSP-EAR, Tnep1
Chromosome Location
Chr.10: 77686030 - 77886994 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287074.1 NP_001274003.1 5 - 6 1036 67
GenBank mRNA AJ487520.1 - 4 - 5 707 67
AK082868.1 - 3 - 4 508 67
JQ815565.1 - 5 - 6 1036 67

View Full Product Details