Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oit1
Entrez Gene ID
18300
Gene Name
oncoprotein induced transcript 1
Gene Aliases
2310076N21Rik, AV067083, EF-7, Fam3d
Chromosome Location
Chr.14: 8348937 - 8378763 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146050.2 NP_666162.1 6 - 7 569 82
GenBank mRNA AK010206.1 - 6 - 7 544 82
BC024504.1 - 6 - 7 552 82
BC024532.1 - 6 - 7 385 82
BC024542.1 - 6 - 7 566 82
BC024636.1 - 1 - 2 56 82
BC034193.1 - 6 - 7 575 82

View Full Product Details