The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110214.1 | NP_001103684.1 | 8 - 9 | 761 | 113 |
NM_146114.3 | NP_666226.2 | 8 - 9 | 761 | 113 | |
NM_175683.4 | NP_783614.1 | 8 - 9 | 761 | 113 | |
GenBank mRNA | AF387731.1 | - | 8 - 9 | 743 | 113 |
AK037126.1 | - | 8 - 9 | 728 | 113 | |
AK038788.1 | - | 8 - 9 | 691 | 113 | |
AK052369.1 | - | 8 - 9 | 717 | 113 | |
AK052490.1 | - | 8 - 9 | 700 | 113 | |
AK088810.1 | - | 6 - 7 | 543 | 113 | |
AK166324.1 | - | 8 - 9 | 704 | 113 | |
BC108935.2 | - | 8 - 9 | 714 | 113 |