The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081456.1 | NP_001074925.1 | 6 - 7 | 945 | 57 |
NM_148937.2 | NP_683739.2 | 6 - 7 | 1096 | 57 | |
GenBank mRNA | AK016945.1 | - | 6 - 7 | 1093 | 57 |
AK039149.1 | - | 6 - 7 | 923 | 57 | |
AK140231.1 | - | 6 - 7 | 932 | 57 | |
AY033991.1 | - | 6 - 7 | 928 | 57 | |
BC066156.1 | - | 6 - 7 | 836 | 57 |