The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081456.1 | NP_001074925.1 | 13 - 14 | 2109 | 105 |
NM_148937.2 | NP_683739.2 | 14 - 15 | 2356 | 105 | |
GenBank mRNA | AK140231.1 | - | 13 - 14 | 2093 | 105 |
AY033991.1 | - | 14 - 15 | 2188 | 105 | |
BC066156.1 | - | 13 - 14 | 2000 | 105 |