Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc8a2
Entrez Gene ID
110891
Gene Name
solute carrier family 8 (sodium/calcium exchanger), member 2
Gene Aliases
Ncx2
Chromosome Location
Chr.7: 16128313 - 16161063 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148946.2 NP_683748.1 6 - 7 2028 93
GenBank mRNA AF503502.1 - 5 - 6 1893 93
AK035163.1 - 6 - 7 2170 93
BC058704.1 - 6 - 7 2028 93

View Full Product Details