The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007485.4 | NP_031511.1 | 1 - 2 | 201 | 70 |
GenBank mRNA | AK008880.1 | - | 1 - 2 | 202 | 70 |
BC005652.1 | - | 1 - 2 | 198 | 70 | |
BC047989.1 | - | 1 - 2 | 210 | 70 | |
D89821.1 | - | - | 212 | 70 | |
X84325.1 | - | 1 - 2 | 128 | 70 |