The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007649.4 | NP_031675.1 | 2 - 3 | 413 | 100 |
GenBank mRNA | AB196820.1 | - | 2 - 3 | 374 | 100 |
AB196821.1 | - | 2 - 3 | 374 | 100 | |
AB196822.1 | - | 2 - 3 | 374 | 100 | |
AB196823.1 | - | 2 - 3 | 374 | 100 | |
AK007741.1 | - | 2 - 3 | 414 | 100 | |
AK084984.1 | - | 2 - 3 | 442 | 100 | |
AK146173.1 | - | 2 - 3 | 460 | 100 | |
AK151601.1 | - | 2 - 3 | 404 | 100 | |
BC060977.1 | - | 2 - 3 | 441 | 100 | |
X17501.1 | - | 2 - 3 | 424 | 100 | |
X53526.1 | - | 2 - 3 | 424 | 100 |