Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dhx9
Entrez Gene ID
13211
Gene Name
DEAH (Asp-Glu-Ala-His) box polypeptide 9
Gene Aliases
AI326842, Ddx9, HEL-5, NDHII, RHA, mHEL-5
Chromosome Location
Chr.1: 153455756 - 153487686 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007842.2 NP_031868.2 16 - 17 1974 71
GenBank mRNA AK135603.1 - 11 - 12 1289 71
AK141824.1 - 16 - 17 1969 71
AY512925.1 - 12 - 13 1430 71
BC089159.1 - 16 - 17 1963 71
U91922.1 - 16 - 17 1931 71
U92080.1 - 5 - 6 669 71

View Full Product Details