The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008236.4 | NP_032262.2 | 4 - 5 | 366 | 62 |
GenBank mRNA | AK053792.1 | - | 4 - 5 | 367 | 62 |
BC138112.1 | - | 4 - 5 | 694 | 62 |