Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc22a1
Entrez Gene ID
20517
Gene Name
solute carrier family 22 (organic cation transporter), member 1
Gene Aliases
Lx1, Oct1, Orct, Orct1
Chromosome Location
Chr.17: 12648874 - 12675838 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009202.5 NP_033228.2 5 - 6 1141 85
GenBank mRNA AF010259.1 - 5 - 6 999 85
BC021651.1 - 5 - 6 1141 85
U38652.1 - 5 - 6 996 85

View Full Product Details