The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009417.2 | NP_033443.1 | 8 - 9 | 1371 | 119 |
GenBank mRNA | AK047843.1 | - | 8 - 9 | 1350 | 119 |
X60703.1 | - | 8 - 9 | 1371 | 119 |