The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009417.2 | NP_033443.1 | 16 - 17 | 2782 | 53 |
GenBank mRNA | AK047843.1 | - | 16 - 17 | 2761 | 53 |
X60703.1 | - | 16 - 17 | 2782 | 53 |