The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009769.4 | NP_033899.2 | 1 - 2 | 535 | 140 |
GenBank mRNA | AB025099.3 | - | 1 - 2 | 509 | 140 |
AF079852.1 | - | 1 - 2 | 397 | 140 | |
AK051409.1 | - | 1 - 2 | 545 | 140 | |
AK087066.1 | - | 1 - 2 | 379 | 141 | |
BC011307.1 | - | 2 - 3 | 353 | 140 | |
BC012958.1 | - | 1 - 2 | 536 | 140 |