Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Frk
Entrez Gene ID
14302
Gene Name
fyn-related kinase
Gene Aliases
BSK, BSK/IYK, C85044, GTK, RAK
Chromosome Location
Chr.10: 34483400 - 34611534 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159544.1 NP_001153016.1 6 - 7 1759 130
NM_010237.3 NP_034367.2 7 - 8 1603 130
GenBank mRNA AK028855.1 - 7 - 8 1543 130
AK052614.1 - 7 - 8 1475 130
AK085191.1 - 6 - 7 1604 130
AK166379.1 - 7 - 8 1603 130
BC007137.1 - 7 - 8 1616 130
L36132.1 - 6 - 7 1638 130
Z48757.1 - 7 - 8 1468 130

View Full Product Details