The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010722.5 | NP_034852.2 | 10 - 11 | 1729 | 128 |
GenBank mRNA | AK132671.1 | - | 7 - 8 | 1437 | 128 |
BC042430.1 | - | 10 - 11 | 1734 | 128 | |
BC051985.1 | - | 10 - 11 | 1705 | 128 | |
D13455.1 | - | 7 - 8 | 1416 | 128 | |
X54098.1 | - | 10 - 11 | 1676 | 128 |