The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010807.4 | NP_034937.1 | 1 - 2 | 338 | 147 |
GenBank mRNA | AK079390.1 | - | 1 - 2 | 287 | 147 |
AK083913.1 | - | 1 - 2 | 289 | 147 | |
AK150261.1 | - | 1 - 2 | 286 | 146 | |
AK152990.1 | - | 1 - 2 | 286 | 147 | |
AK169355.1 | - | 1 - 2 | 289 | 147 | |
BC006757.1 | - | 1 - 2 | 266 | 147 | |
CX243729.1 | - | 1 - 2 | 338 | 147 | |
X61399.1 | - | 1 - 2 | 275 | 147 |