The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010893.3 | NP_035023.3 | 2 - 3 | 487 | 123 |
GenBank mRNA | AK077444.1 | - | 2 - 3 | 427 | 123 |
AK145680.1 | - | 2 - 3 | 381 | 123 | |
BC004666.1 | - | 2 - 3 | 361 | 123 | |
Y11412.1 | - | 2 - 3 | 366 | 123 |