The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011486.4 | NP_035616.1 | 4 - 5 | 648 | 93 |
NM_213659.2 | NP_998824.1 | 4 - 5 | 648 | 93 | |
NM_213660.2 | NP_998825.1 | 4 - 5 | 648 | 93 | |
GenBank mRNA | AK145486.1 | - | 4 - 5 | 504 | 93 |
AK153005.1 | - | 4 - 5 | 560 | 93 | |
AK153472.1 | - | 4 - 5 | 560 | 93 | |
AY299489.1 | - | 3 - 4 | 375 | 93 | |
AY299490.1 | - | 3 - 4 | 375 | 93 | |
BC003806.1 | - | 4 - 5 | 536 | 93 | |
BC019168.1 | - | 4 - 5 | 552 | 93 | |
U06922.1 | - | 4 - 5 | 437 | 93 | |
U08378.1 | - | 4 - 5 | 515 | 93 | |
U30709.1 | - | 4 - 5 | 541 | 93 |