Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vill
Entrez Gene ID
22351
Gene Name
villin-like
Gene Aliases
Villp
Chromosome Location
Chr.9: 119052729 - 119071525 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164567.1 NP_001158039.1 8 - 9 906 88
NM_011700.2 NP_035830.2 8 - 9 906 88
GenBank mRNA AJ344341.1 - 8 - 9 894 88
AK028229.1 - 8 - 9 907 88
AK166823.1 - 7 - 8 1138 88
BC021808.1 - 8 - 9 882 88
CF171710.1 - 4 - 5 463 88

View Full Product Details