The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164567.1 | NP_001158039.1 | 11 - 12 | 1240 | 74 |
NM_011700.2 | NP_035830.2 | 11 - 12 | 1240 | 74 | |
GenBank mRNA | AJ344341.1 | - | 11 - 12 | 1228 | 74 |
AK028229.1 | - | 11 - 12 | 1241 | 74 | |
AK166823.1 | - | 10 - 11 | 1472 | 74 | |
BC021808.1 | - | 11 - 12 | 1216 | 74 | |
BC022664.1 | - | 2 - 3 | 169 | 74 |