The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011959.2 | NP_036089.1 | 6 - 7 | 756 | 90 |
GenBank mRNA | AJ007360.1 | - | 6 - 7 | 753 | 90 |
AK084683.1 | - | 6 - 7 | 784 | 90 | |
AK165912.1 | - | 6 - 7 | 762 | 90 | |
AK166537.1 | - | 6 - 7 | 695 | 90 | |
BC006927.1 | - | 6 - 7 | 772 | 90 |