Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clasrp
Entrez Gene ID
53609
Gene Name
CLK4-associating serine/arginine rich protein
Gene Aliases
Clasp, Sfrs16, Srsf16, Swap2
Chromosome Location
Chr.7: 19581038 - 19604517 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016680.5 NP_057889.3 10 - 11 952 85
NR_125832.1 10 - 11 952 85
NR_125833.1 10 - 11 936 85
NR_125834.1 10 - 11 952 85
NR_125835.1 10 - 11 914 85
NR_125836.1 8 - 9 816 85
GenBank mRNA AB080582.1 - 9 - 10 815 85
AB080583.1 - 9 - 10 815 85
AF042799.1 - 10 - 11 917 85
AK009334.1 - 10 - 11 903 85
AK159443.1 - 10 - 11 921 85
BC010728.1 - 10 - 11 903 85
BC051916.1 - 10 - 11 927 85
BC060092.1 - 9 - 10 3320 85
BC079543.1 - 9 - 10 3310 85
BC103781.1 - 8 - 9 782 85
BC106129.1 - 10 - 11 915 85

View Full Product Details