Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clasrp
Entrez Gene ID
53609
Gene Name
CLK4-associating serine/arginine rich protein
Gene Aliases
Clasp, Sfrs16, Srsf16, Swap2
Chromosome Location
Chr.7: 19581038 - 19604517 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016680.5 NP_057889.3 16 - 17 1848 87
NR_125832.1 16 - 17 1852 87
NR_125833.1 15 - 16 2085 87
NR_125834.1 15 - 16 2101 87
NR_125835.1 15 - 16 2063 87
NR_125836.1 13 - 14 1965 87
GenBank mRNA AB080582.1 - 15 - 16 1711 87
AB080583.1 - 15 - 16 1715 87
AF042799.1 - 15 - 16 2066 87
AF325456.1 - 6 - 7 1694 87
AK009334.1 - 15 - 16 2052 87
AK159443.1 - 16 - 17 1821 87
BC010728.1 - 16 - 17 1800 87
BC037756.1 - 1 - 2 185 87
BC051916.1 - 16 - 17 1827 87
BC060092.1 - 15 - 16 4216 87
BC103781.1 - 13 - 14 1931 87
BC106129.1 - 15 - 16 2064 87

View Full Product Details