Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdyn
Entrez Gene ID
18610
Gene Name
prodynorphin
Gene Aliases
Dyn
Chromosome Location
Chr.2: 129686549 - 129699938 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286502.1 NP_001273431.1 2 - 3 103 82
NM_018863.4 NP_061351.2 2 - 3 294 82
GenBank mRNA AF026537.1 - 2 - 3 189 82
AK133597.1 - 2 - 3 295 82
BC137571.1 - 2 - 3 133 82
BC137572.1 - 2 - 3 133 82
BY292284.1 - 2 - 3 104 82

View Full Product Details