The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161428.1 | NP_001154900.1 | 1 - 2 | 168 | 84 |
NM_019449.2 | NP_062322.2 | 1 - 2 | 168 | 84 | |
GenBank mRNA | AK030488.1 | - | 1 - 2 | 129 | 84 |
AK039526.1 | - | 1 - 2 | 129 | 84 | |
AK155556.1 | - | 1 - 2 | 126 | 84 | |
AK162535.1 | - | 1 - 2 | 121 | 84 | |
AK165060.1 | - | 1 - 2 | 128 | 84 | |
BC018388.1 | - | 1 - 2 | 135 | 84 | |
BY199008.1 | - | 1 - 2 | 168 | 84 |