The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021557.5 | NP_067532.2 | 1 - 2 | 94 | 88 |
GenBank mRNA | AB035959.1 | - | 1 - 2 | 102 | 88 |
AF474027.1 | - | 1 - 2 | 140 | 88 | |
AK004413.1 | - | 1 - 2 | 95 | 88 | |
AK012056.1 | - | 1 - 2 | 122 | 88 | |
AK135443.1 | - | 1 - 2 | 124 | 88 | |
AY039032.1 | - | 1 - 2 | 568 | 88 | |
BC018261.1 | - | 1 - 2 | 119 | 88 | |
BC036273.1 | - | 1 - 2 | 119 | 88 |