The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199105.1 | NP_001186034.1 | 2 - 3 | 373 | 97 |
NM_021897.3 | NP_068697.1 | 2 - 3 | 373 | 97 | |
GenBank mRNA | AJ131776.1 | - | 2 - 3 | 374 | 97 |
AK142321.1 | - | 2 - 3 | 369 | 97 | |
AY034611.1 | - | 2 - 3 | 374 | 97 | |
AY034612.1 | - | 2 - 3 | 374 | 97 | |
BC132124.1 | - | 1 - 2 | 275 | 97 | |
BC138711.1 | - | 1 - 2 | 275 | 97 | |
BC145396.1 | - | 1 - 2 | 274 | 97 |