The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195774.1 | NP_001182703.1 | 1 - 2 | 173 | 89 |
NM_022420.2 | NP_071865.1 | 1 - 2 | 173 | 89 | |
GenBank mRNA | AF378831.1 | - | 1 - 2 | 125 | 89 |
BC020004.1 | - | 1 - 2 | 124 | 89 | |
CJ054496.1 | - | 1 - 2 | 174 | 89 |