The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022422.5 | NP_071867.1 | 2 - 3 | 239 | 92 |
GenBank mRNA | AB030194.1 | - | 2 - 3 | 188 | 92 |
AK018778.1 | - | 2 - 3 | 188 | 92 | |
AY029485.1 | - | 2 - 3 | 188 | 92 | |
BC048431.1 | - | 2 - 3 | 187 | 92 | |
BC059712.1 | - | 2 - 3 | 186 | 92 | |
BU554843.1 | - | 2 - 3 | 243 | 92 | |
EH103048.1 | - | - | 66 | 92 |