The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022997.4 | NP_075373.1 | 2 - 3 | 141 | 65 |
GenBank mRNA | AF226323.1 | - | 2 - 3 | 160 | 65 |
AK135680.1 | - | 2 - 3 | 447 | 65 | |
AK142382.1 | - | 2 - 3 | 428 | 65 | |
AK145421.1 | - | 1 - 2 | 108 | 65 | |
AK145853.1 | - | 2 - 3 | 191 | 65 | |
AK150682.1 | - | 2 - 3 | 155 | 65 | |
AK160547.1 | - | 1 - 2 | 112 | 65 | |
AK162726.1 | - | 2 - 3 | 120 | 65 | |
AK166821.1 | - | 2 - 3 | 134 | 65 | |
AK167421.1 | - | 2 - 3 | 158 | 65 | |
AK167594.1 | - | 2 - 3 | 154 | 65 | |
AK168880.1 | - | 2 - 3 | 134 | 65 | |
AK170473.1 | - | 2 - 3 | 175 | 65 | |
BC005469.1 | - | 2 - 3 | 166 | 65 | |
BC006637.1 | - | 2 - 3 | 146 | 65 | |
U47024.1 | - | 1 - 2 | 105 | 65 |