The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023196.4 | NP_075685.2 | 3 - 4 | 570 | 58 |
NM_183423.3 | NP_904359.1 | 3 - 4 | 353 | 58 | |
GenBank mRNA | AK003183.1 | - | 3 - 4 | 530 | 58 |
AK009133.1 | - | 3 - 4 | 572 | 58 | |
AK010011.1 | - | 3 - 4 | 560 | 58 | |
AK010174.1 | - | 3 - 4 | 536 | 58 | |
AK012414.1 | - | 3 - 4 | 498 | 58 | |
AK163693.1 | - | 3 - 4 | 484 | 58 | |
AY007381.1 | - | 3 - 4 | 543 | 58 | |
AY007382.1 | - | 3 - 4 | 353 | 58 | |
BC026812.1 | - | 3 - 4 | 535 | 58 | |
BC051117.1 | - | 3 - 4 | 570 | 58 |