The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023463.3 | NP_075952.1 | 2 - 3 | 233 | 63 |
GenBank mRNA | AJ315546.1 | - | 2 - 3 | 168 | 63 |
AK003375.1 | - | 2 - 3 | 234 | 63 | |
BC116365.1 | - | 2 - 3 | 199 | 63 | |
BC116366.1 | - | 2 - 3 | 197 | 63 |