Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ripk4
Entrez Gene ID
72388
Gene Name
receptor-interacting serine-threonine kinase 4
Gene Aliases
2310069J12Rik, AI552420, ANKK2, Ankrd3, DIk, PKK, RIP4
Chromosome Location
Chr.16: 97741933 - 97763755 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023663.6 NP_076152.2 2 - 3 521 84
GenBank mRNA AF302127.3 - 2 - 3 524 84
AK077233.1 - 2 - 3 706 84
AK145203.1 - 2 - 3 503 84
BC057871.1 - 2 - 3 484 84

View Full Product Details