Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ripk4
Entrez Gene ID
72388
Gene Name
receptor-interacting serine-threonine kinase 4
Gene Aliases
2310069J12Rik, AI552420, ANKK2, Ankrd3, DIk, PKK, RIP4
Chromosome Location
Chr.16: 97741933 - 97763755 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023663.6 NP_076152.2 6 - 7 978 75
GenBank mRNA AF302127.3 - 6 - 7 981 75
AK077233.1 - 6 - 7 1162 75
AK145203.1 - 6 - 7 960 75
BC057871.1 - 6 - 7 941 75

View Full Product Details