Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep70
Entrez Gene ID
68121
Gene Name
centrosomal protein 70
Gene Aliases
6720484E09Rik, Bite, C030018L16Rik
Chromosome Location
Chr.9: 99183595 - 99302065 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023873.3 NP_076362.2 7 - 8 813 113
GenBank mRNA AF242319.1 - 7 - 8 849 113
AK021095.1 - 7 - 8 944 113
AK133048.1 - 7 - 8 819 113
BC050819.1 - 7 - 8 805 113
BC071260.1 - 7 - 8 813 113

View Full Product Details