The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024222.2 | NP_077184.2 | 13 - 14 | 2423 | 120 |
GenBank mRNA | AK012153.1 | - | 13 - 14 | 2093 | 120 |
AK018758.2 | - | 13 - 14 | 2133 | 120 | |
AK145674.1 | - | 13 - 14 | 2064 | 120 | |
AK148065.1 | - | 13 - 14 | 2122 | 120 | |
AK152899.1 | - | 13 - 14 | 2471 | 120 | |
AK154979.1 | - | 13 - 14 | 2112 | 120 | |
AK170079.1 | - | 13 - 14 | 2135 | 120 | |
AX799084.1 | - | 13 - 14 | 2134 | 120 | |
BC003206.1 | - | 8 - 9 | 1135 | 120 | |
BC013054.1 | - | 3 - 4 | 541 | 120 | |
BC052433.1 | - | 13 - 14 | 2423 | 120 |