Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Stt3b
- Entrez Gene ID
-
68292
- Gene Name
- STT3, subunit of the oligosaccharyltransferase complex, homolog B (S. cerevisiae)
- Gene Aliases
-
1300006C19Rik, Simp
- Chromosome Location
-
Chr.9: 115242581 - 115310421 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC013054.1 |
- |
4 - 5 |
665 |
111 |
View Full Product Details