The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025576.2 | NP_079852.1 | 2 - 3 | 519 | 64 |
GenBank mRNA | AK012674.1 | - | 2 - 3 | 509 | 64 |
AK014691.1 | - | 2 - 3 | 513 | 64 | |
AK151972.1 | - | 2 - 3 | 520 | 64 | |
BC026750.1 | - | 2 - 3 | 498 | 64 |