The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025576.2 | NP_079852.1 | 3 - 4 | 712 | 88 |
GenBank mRNA | AK003210.1 | - | 3 - 4 | 416 | 88 |
AK012674.1 | - | 3 - 4 | 702 | 88 | |
AK014691.1 | - | 3 - 4 | 706 | 88 | |
AK151972.1 | - | 3 - 4 | 713 | 88 | |
AK168416.1 | - | - | 578 | 88 | |
BC026750.1 | - | 3 - 4 | 691 | 88 |