The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025638.2 | NP_079914.1 | 2 - 3 | 346 | 73 |
GenBank mRNA | AK013864.1 | - | 2 - 3 | 272 | 73 |
AK016023.1 | - | 2 - 3 | 272 | 73 | |
AK167812.1 | - | 2 - 3 | 329 | 73 | |
BC016541.1 | - | 2 - 3 | 269 | 73 | |
BY282485.1 | - | 2 - 3 | 347 | 73 |