The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025696.3 | NP_079972.1 | 5 - 6 | 1173 | 54 |
GenBank mRNA | AF276314.1 | - | 5 - 6 | 1193 | 54 |
AK158805.1 | - | 5 - 6 | 1041 | 54 |