Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map1lc3a
Entrez Gene ID
66734
Gene Name
microtubule-associated protein 1 light chain 3 alpha
Gene Aliases
1010001H21Rik, 4922501H04Rik, LC3, LC3a
Chromosome Location
Chr.2: 155276364 - 155278073 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025735.3 NP_080011.1 2 - 3 334 74
GenBank mRNA AK003122.1 - 2 - 3 245 74
AK003716.1 - 2 - 3 243 74
AK015020.1 - 2 - 3 253 74
BC010596.1 - 2 - 3 234 74
CA322683.1 - 2 - 3 403 74

View Full Product Details