The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284332.1 | NP_001271261.1 | 4 - 5 | 561 | 69 |
NM_025858.3 | NP_080134.1 | 4 - 5 | 558 | 69 | |
NM_026381.5 | NP_080657.1 | 2 - 3 | 258 | 69 | |
GenBank mRNA | AF520699.1 | - | 4 - 5 | 540 | 69 |
AK009225.1 | - | 4 - 5 | 498 | 69 | |
AK018291.1 | - | 2 - 3 | 214 | 69 | |
AK020877.1 | - | 2 - 3 | 235 | 69 | |
AK033545.1 | - | 4 - 5 | 518 | 69 | |
AK045368.1 | - | 4 - 5 | 488 | 69 | |
AK081060.1 | - | 2 - 3 | 236 | 69 | |
AK088188.1 | - | 3 - 4 | 254 | 69 | |
AK090357.1 | - | 2 - 3 | 412 | 69 | |
AK090373.1 | - | 4 - 5 | 497 | 69 | |
AK205406.1 | - | 1 - 2 | 130 | 69 | |
AK212074.1 | - | 2 - 3 | 217 | 69 | |
BC010238.1 | - | 4 - 5 | 522 | 69 | |
BC017600.1 | - | 2 - 3 | 170 | 69 | |
BC057089.1 | - | 2 - 3 | 210 | 69 |