Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdcl
Entrez Gene ID
67466
Gene Name
phosducin-like
Gene Aliases
1200011E13Rik, AW108059, PhLP1
Chromosome Location
Chr.2: 37350074 - 37359820 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026176.3 NP_080452.2 1 - 2 190 75
GenBank mRNA AK028668.1 - 1 - 2 80 75
AK028814.1 - 1 - 2 80 75
AK133747.1 - 1 - 2 100 75
AK166983.1 - 1 - 2 80 75
BC006578.1 - 1 - 2 90 75
BY024444.1 - 1 - 2 190 75

View Full Product Details