The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026918.2 | NP_081194.1 | 2 - 3 | 192 | 65 |
GenBank mRNA | BC031800.1 | - | 2 - 3 | 192 | 65 |